Assay Details
| Gene Symbol: LOC111094603 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_022415442.1 | XP_022271150.1 | 2 - 3 | 667 | 84 | |
| XM_022415443.1 | XP_022271151.1 | 2 - 3 | 670 | 84 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_022415442.1, XM_022415443.1 transcripts.
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

LOC111094603
Genomic Map