Assay Details
| Gene Symbol: CHMP4A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_014115792.1 | XP_013971267.1 | 1 - 2 | 173 | 149 | |
| XM_537387.5 | XP_537387.3 | 1 - 2 | 173 | 149 | ||
More Information
Set Membership: |
|

CHMP4A
Genomic Map