Assay Details
| Gene Symbol: SUMO1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001252263.1 | NP_001239192.1 | 1 - 2 | 108 | 70 | |
| GenBank mRNA | BQ290300.1 | - | 1 - 2 | 119 | 70 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001252263.1 transcripts.
Set Membership: |
|

SUMO1
Genomic Map