Assay Details
| Gene Symbol: CTH | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_022420631.1 | XP_022276339.1 | 5 - 6 | 776 | 127 | |
| XM_537115.6 | XP_537115.2 | 5 - 6 | 927 | 127 | ||
More Information
Set Membership: |
|

CTH
Genomic Map