Assay Details
| Gene Symbol: GPC3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_022415889.1 | XP_022271597.1 | 8 - 9 | 1826 | 79 | |
| XM_538178.6 | XP_538178.2 | 7 - 8 | 1571 | 79 | ||
More Information
Set Membership: |
|

GPC3
Genomic Map