Assay Details
| Gene Symbol: LAMTOR2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005622713.3 | XP_005622770.1 | 4 - 5 | 532 | 58 | |
| XM_022421275.1 | XP_022276983.1 | 4 - 5 | 570 | 58 | ||
| XM_537247.6 | XP_537247.1 | 3 - 4 | 456 | 58 | ||
More Information
Set Membership: |
|

LAMTOR2
Genomic Map