Assay Details
| Gene Symbol: PIGL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005620092.3 | XP_005620149.1 | 1 - 2 | 913 | 81 | |
| XM_014113621.2 | XP_013969096.1 | 1 - 2 | 913 | 81 | ||
| XM_022418623.1 | XP_022274331.1 | 2 - 3 | 928 | 81 | ||
More Information
Set Membership: |
|

PIGL
Genomic Map