Assay Details
| Gene Symbol: MMD | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_022423162.1 | XP_022278870.1 | 337 | 77 | ||
| XM_022423163.1 | XP_022278871.1 | 337 | 77 | |||
| XM_537684.6 | XP_537684.1 | 337 | 77 | |||
More Information
Set Membership: |
|

MMD
Genomic Map