Assay Details
| Gene Symbol: LOC480667 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_537786.6 | XP_537786.2 | 8 - 9 | 1251 | 71 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC480667| Gene Symbol: LOC480667 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_537786.6 | XP_537786.2 | 8 - 9 | 1251 | 71 | |
Set Membership: |
|