Assay Details
| Gene Symbol: HCCS | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005641099.3 | XP_005641156.1 | 2 - 3 | 298 | 78 | |
| XM_005641100.3 | XP_005641157.1 | 2 - 3 | 333 | 78 | ||
More Information
Set Membership: |
|

HCCS
Genomic Map