Assay Details
| Gene Symbol: CENPN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_014113992.2 | XP_013969467.1 | 2 - 3 | 368 | 67 | |
| XM_536770.5 | XP_536770.3 | 2 - 3 | 335 | 67 | ||
More Information
Set Membership: |
|

CENPN
Genomic Map