Assay Details
| Gene Symbol: LOC100856583 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003639354.2 | XP_003639402.1 | 1 | 371 | 98 | |
| Gene Symbol: ARL6IP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_537908.7 | XP_537908.4 | 3 - 4 | 409 | 98 | |
More Information
Set Membership: |
|

LOC100856583
Genomic Map