Assay Details
| Gene Symbol: DYNLRB2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_536771.6 | XP_536771.1 | 2 - 3 | 218 | 82 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

DYNLRB2| Gene Symbol: DYNLRB2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_536771.6 | XP_536771.1 | 2 - 3 | 218 | 82 | |
Set Membership: |
|