Assay Details
| Gene Symbol: LOC479822 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005621477.2 | XP_005621534.1 | 9 - 10 | 1294 | 98 | |
| XM_005621478.2 | XP_005621535.1 | 7 - 8 | 1057 | 98 | ||
| XM_536949.5 | XP_536949.3 | 9 - 10 | 1294 | 98 | ||
More Information
Set Membership: |
|

LOC479822
Genomic Map