Assay Details
| Gene Symbol: LOC479822 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005621477.2 | XP_005621534.1 | 10 - 11 | 1397 | 91 | |
| XM_005621478.2 | XP_005621535.1 | 8 - 9 | 1160 | 91 | ||
| XM_536949.5 | XP_536949.3 | 10 - 11 | 1397 | 91 | ||
More Information
Set Membership: |
|

LOC479822
Genomic Map