Assay Details
| Gene Symbol: LOC479822 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005621478.2 | XP_005621535.1 | 9 - 10 | 1285 | 93 | |
| XM_536949.5 | XP_536949.3 | 11 - 12 | 1522 | 93 | ||
More Information
Set Membership: |
|

LOC479822
Genomic Map