Assay Details
| Gene Symbol: TMEM218 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003434543.4 | XP_003434591.1 | 360 | 59 | ||
| XM_536536.6 | XP_536536.1 | 358 | 59 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

TMEM218| Gene Symbol: TMEM218 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003434543.4 | XP_003434591.1 | 360 | 59 | ||
| XM_536536.6 | XP_536536.1 | 358 | 59 | |||
Set Membership: |
|