Assay Details
| Gene Symbol: SPSB1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_022419028.1 | XP_022274736.1 | 1057 | 64 | ||
| XM_022419029.1 | XP_022274737.1 | 1058 | 64 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: SPSB1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_022419028.1 | XP_022274736.1 | 1057 | 64 | ||
| XM_022419029.1 | XP_022274737.1 | 1058 | 64 | |||
Set Membership: |
|