Assay Details
Gene Symbol: ZNF25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005618772.3 | XP_005618829.1 | 468 | 90 | ||
XM_005618773.3 | XP_005618830.1 | 635 | 90 | |||
XM_014112717.2 | XP_013968192.1 | 348 | 90 | |||
XM_022417372.1 | XP_022273080.1 | 468 | 90 | |||
XM_536326.7 | XP_536326.5 | 519 | 90 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
