Assay Details
| Gene Symbol: AK1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001131047.1 | NP_001124519.1 | 3 - 4 | 312 | 72 | |
| XM_005625019.2 | XP_005625076.1 | 4 - 5 | 362 | 72 | ||
More Information
Set Membership: |
|

AK1
Genomic Map