Assay Details
| Gene Symbol: AIMP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005621115.3 | XP_005621172.1 | 362 | 72 | ||
| XM_022419726.1 | XP_022275434.1 | 342 | 72 | |||
| XM_022419727.1 | XP_022275435.1 | 333 | 72 | |||
| XM_536880.6 | XP_536880.2 | 482 | 72 | |||
More Information
Set Membership: |
|

AIMP2
Genomic Map