Assay Details
Gene Symbol: SPRYD4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_538234.4 | XP_538234.1 | 176 | 145 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
