Assay Details
| Gene Symbol: GPKOW | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005641349.3 | XP_005641406.1 | 4 - 5 | 843 | 103 | |
| XM_005641350.2 | XP_005641407.1 | 4 - 5 | 833 | 103 | ||
| XM_014111765.2 | XP_013967240.1 | 4 - 5 | 847 | 103 | ||
| XM_014111766.2 | XP_013967241.1 | 4 - 5 | 748 | 103 | ||
More Information
Set Membership: |
|

GPKOW
Genomic Map