Assay Details
| Gene Symbol: LOC479708 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005620930.3 | XP_005620987.1 | 1695 | 73 | ||
| XM_022419526.1 | XP_022275234.1 | 1696 | 73 | |||
| XM_022419527.1 | XP_022275235.1 | 1696 | 73 | |||
More Information
Set Membership: |
|

LOC479708
Genomic Map