Assay Details
| Gene Symbol: SUN3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_022405136.1 | XP_022260844.1 | 861 | 88 | ||
| XM_022405137.1 | XP_022260845.1 | 860 | 88 | |||
| XM_022405139.1 | XP_022260847.1 | 859 | 88 | |||
| XM_537884.6 | XP_537884.4 | 859 | 88 | |||
More Information
Set Membership: |
|

SUN3
Genomic Map