Assay Details
| Gene Symbol: CMTM2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005620875.2 | XP_005620932.1 | 2 - 3 | 570 | 112 | |
| XM_536824.5 | XP_536824.3 | 2 - 3 | 565 | 112 | ||
More Information
Set Membership: |
|

CMTM2
Genomic Map