Assay Details
| Gene Symbol: LOC480424 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005623771.3 | XP_005623828.1 | 3 - 4 | 1018 | 71 | |
| XM_022422352.1 | XP_022278060.1 | 3 - 4 | 1266 | 71 | ||
| XM_537545.5 | XP_537545.1 | 3 - 4 | 1051 | 71 | ||
More Information
Set Membership: |
|

LOC480424
Genomic Map