Assay Details
| Gene Symbol: ZFAND2A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_022419811.1 | XP_022275519.1 | 578 | 76 | ||
| XM_537924.6 | XP_537924.2 | 569 | 76 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

ZFAND2A| Gene Symbol: ZFAND2A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_022419811.1 | XP_022275519.1 | 578 | 76 | ||
| XM_537924.6 | XP_537924.2 | 569 | 76 | |||
Set Membership: |
|