Assay Details
| Gene Symbol: DNASE2B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_022420577.1 | XP_022276285.1 | 1 - 2 | 354 | 84 | |
| XM_022420578.1 | XP_022276286.1 | 1 - 2 | 354 | 84 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_022420578.1 transcripts.
Set Membership: |
|

DNASE2B
Genomic Map