Assay Details
| Gene Symbol: CCNH | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005618142.3 | XP_005618199.1 | 901 | 96 | ||
| XM_536300.6 | XP_536300.3 | 901 | 96 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: CCNH | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005618142.3 | XP_005618199.1 | 901 | 96 | ||
| XM_536300.6 | XP_536300.3 | 901 | 96 | |||
Set Membership: |
|