Assay Details
| Gene Symbol: ENTHD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_014117347.2 | XP_013972822.1 | 2 - 3 | 618 | 129 | |
| XM_022424398.1 | XP_022280106.1 | 3 - 4 | 993 | 129 | ||
| XM_538363.6 | XP_538363.4 | 2 - 3 | 618 | 129 | ||
More Information
Set Membership: |
|

ENTHD1
Genomic Map