Assay Details
| Gene Symbol: ILRUN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003431707.3 | XP_003431755.1 | 1 - 2 | 354 | 57 | |
| XM_022425952.1 | XP_022281660.1 | 1 - 2 | 362 | 57 | ||
More Information
Set Membership: |
|

ILRUN
Genomic Map