Assay Details
| Gene Symbol: ORC6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_014119635.2 | XP_013975110.1 | 4 - 5 | 1080 | 75 | |
| XM_014119636.2 | XP_013975111.1 | 4 - 5 | 937 | 75 | ||
| XM_014119638.2 | XP_013975113.1 | 2 - 3 | 714 | 75 | ||
| XM_022427882.1 | XP_022283590.1 | 4 - 5 | 723 | 75 | ||
| XM_022427883.1 | XP_022283591.1 | 4 - 5 | 582 | 75 | ||
| XM_539609.5 | XP_539609.3 | 3 - 4 | 834 | 75 | ||
More Information
Set Membership: |
|

ORC6
Genomic Map