Assay Details
| Gene Symbol: LOC611720 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_022422175.1 | XP_022277883.1 | 2 - 3 | 1214 | 103 | |
| XM_022422176.1 | XP_022277884.1 | 2 - 3 | 1214 | 103 | ||
| XM_022422178.1 | XP_022277886.1 | 4 - 5 | 402 | 103 | ||
| Gene Symbol: NUDT6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005631832.3 | XP_005631889.1 | 1 - 2 | 149 | 103 | |
| XM_014121452.2 | XP_013976927.1 | 628 | 103 | |||
| XM_022405963.1 | XP_022261671.1 | 340 | 103 | |||
More Information
Set Membership: |
|

LOC611720
Genomic Map