Assay Details
Gene Symbol: NAT9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005624120.3 | XP_005624177.1 | 470 | 70 | ||
XM_005624121.3 | XP_005624178.1 | 472 | 70 | |||
XM_014116308.2 | XP_013971783.1 | 480 | 70 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
