Assay Details
| Gene Symbol: SEPTIN14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_022405269.1 | XP_022260977.1 | 266 | 88 | ||
| XM_022405270.1 | XP_022260978.1 | 203 | 88 | |||
| XM_022405271.1 | XP_022260979.1 | 275 | 88 | |||
| XM_540501.5 | XP_540501.2 | 319 | 88 | |||
More Information
Set Membership: |
|

SEPTIN14
Genomic Map