Assay Details
Gene Symbol: TRAPPC5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005632939.3 | XP_005632996.1 | 2 - 3 | 137 | 66 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_005632939.3 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
