Assay Details
| Gene Symbol: METTL25 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_022428133.1 | XP_022283841.1 | 10 - 11 | 1685 | 102 | |
| XM_022428134.1 | XP_022283842.1 | 10 - 11 | 1685 | 102 | ||
More Information
Set Membership: |
|

METTL25
Genomic Map