Assay Details
| Gene Symbol: LOC484867 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_014122176.1 | XP_013977651.1 | 10 - 11 | 1395 | 83 | |
| XM_014122177.2 | XP_013977652.1 | 10 - 11 | 1402 | 83 | ||
| XM_014122178.2 | XP_013977653.1 | 10 - 11 | 1351 | 83 | ||
| XM_014122179.2 | XP_013977654.1 | 10 - 11 | 1388 | 83 | ||
More Information
Set Membership: |
|

LOC484867
Genomic Map