Assay Details
| Gene Symbol: NAGA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005625812.3 | XP_005625869.1 | 7 - 8 | 969 | 70 | |
| XM_005625813.3 | XP_005625870.1 | 7 - 8 | 969 | 70 | ||
| XM_538347.5 | XP_538347.2 | 7 - 8 | 943 | 70 | ||
More Information
Set Membership: |
|

NAGA
Genomic Map