Assay Details
| Gene Symbol: LOC481722 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_022425780.1 | XP_022281488.1 | 35 - 36 | 4797 | 74 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC481722| Gene Symbol: LOC481722 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_022425780.1 | XP_022281488.1 | 35 - 36 | 4797 | 74 | |
Set Membership: |
|