Assay Details
| Gene Symbol: PRAG1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_022404114.1 | XP_022259822.1 | 2170 | 100 | ||
| XM_540000.4 | XP_540000.3 | 2366 | 100 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: PRAG1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_022404114.1 | XP_022259822.1 | 2170 | 100 | ||
| XM_540000.4 | XP_540000.3 | 2366 | 100 | |||
Set Membership: |
|