Assay Details
| Gene Symbol: LOC484344 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005616250.3 | XP_005616307.1 | 4 - 5 | 837 | 87 | |
| XM_014115019.2 | XP_013970494.1 | 4 - 5 | 837 | 87 | ||
More Information
Set Membership: |
|

LOC484344
Genomic Map