Assay Details
| Gene Symbol: CSF2RB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005625909.2 | XP_005625966.1 | 7 - 8 | 1026 | 70 | |
| XM_538397.6 | XP_538397.3 | 10 - 11 | 1624 | 70 | ||
More Information
Set Membership: |
|

CSF2RB
Genomic Map