Assay Details
| Gene Symbol: LOC484867 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005632760.3 | XP_005632817.1 | 3 - 4 | 468 | 93 | |
| XM_014122175.2 | XP_013977650.1 | 2 - 3 | 591 | 93 | ||
| XM_014122180.2 | XP_013977655.1 | 3 - 4 | 488 | 93 | ||
| XM_022406939.1 | XP_022262647.1 | 3 - 4 | 204 | 93 | ||
| XM_541983.6 | XP_541983.3 | 3 - 4 | 488 | 93 | ||
More Information
Set Membership: |
|

LOC484867
Genomic Map