Assay Details
| Gene Symbol: RCN3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_022423812.1 | XP_022279520.1 | 6 - 7 | 1072 | 65 | |
| XM_541494.6 | XP_541494.2 | 6 - 7 | 1081 | 65 | ||
More Information
Set Membership: |
|

RCN3
Genomic Map