Assay Details
| Gene Symbol: ABHD14A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005632467.3 | XP_005632524.1 | 5 - 6 | 929 | 70 | |
| XM_005632470.1 | XP_005632527.1 | 4 - 5 | 689 | 70 | ||
| XM_005632471.2 | XP_005632528.1 | 5 - 6 | 751 | 70 | ||
| XM_541859.6 | XP_541859.4 | 4 - 5 | 1416 | 70 | ||
More Information
Set Membership: |
|

ABHD14A
Genomic Map