Assay Details
| Gene Symbol: SEPTIN9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_022423896.1 | XP_022279604.1 | 1416 | 71 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

SEPTIN9| Gene Symbol: SEPTIN9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_022423896.1 | XP_022279604.1 | 1416 | 71 | ||
Set Membership: |
|