Assay Details
| Gene Symbol: LOC481187 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005625705.3 | XP_005625762.1 | 2 - 3 | 348 | 58 | |
| XM_022424249.1 | XP_022279957.1 | 3 - 4 | 762 | 58 | ||
| XM_538308.6 | XP_538308.5 | 2 - 3 | 1063 | 58 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_005625705.3, XM_022424249.1, XM_538308.6 transcripts.
Set Membership: |
|

LOC481187
Genomic Map