Assay Details
| Gene Symbol: MRPS12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005616606.3 | XP_005616663.1 | 2 - 3 | 159 | 98 | |
| XM_022425830.1 | XP_022281538.1 | 2 - 3 | 152 | 98 | ||
| XM_541631.6 | XP_541631.3 | 2 - 3 | 152 | 98 | ||
More Information
Set Membership: |
|

MRPS12
Genomic Map